missing translation for 'onlineSavingsMsg'
Learn More

Hammond™ Drierite™ Desiccant Bags, Cole-Parmer™

Catalog Number 11833092
Click to view available options
Quantity:
3 g
6 g
1 oz.
8 oz.
This item is not returnable. View return policy
This item is not returnable. View return policy

Protect materials from corrosion with these desiccant bags. Hammond™ Drierite™ Desiccant Bags reduce the humidity to a 100°F dew point in 10 hours and are dust-tight. Materials packaged with these bags are protected from corrosion for many months.

Nonwoven crepe bags filled with Drierite (anyhydrous calcium sulfate) completely dry the air within a package or container during shipment or storage.
  • Sturdy and held together by heat seals
  • Perform well in elevated temperatures
  • Will not release absorbed moisture at less than 350°F
Material Calcium Sulfate
Quantity 8 oz.
Suitability These Bags are Dust-Tight and Will Protect For Many Months
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Drierite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H400-Very toxic to aquatic life.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : calcium sulphate, cobalt dichloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'