missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PTFE Tubing Adapted to Femaile Luer Lock

Connect to a syringe or other component

Brand:  Hamilton™ 90619

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10642503

  • £38.45 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Made of PTFE
  • One female luer fitting on the end
  • Tubing is 12 in. in length

Specifications

Specifications

19 ga.
30.48cm
Tubing Assembly
Connect to a syringe or other component
PTFE
Videos