Learn More
Hamilton™ PRP™Series Guard Cartridge Replacement Cartridges
Replacement Cartridges for PRP-1, PRP-3, PRP-X100, PRP-X200, PRP-X300, PRP-X400
Brand:  Hamilton™ 79446
Code : 16
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10686591
  • £351.00 / Pack of 5
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
Stainless steel or PEEK replacement cartridges.
Specifications
Specifications
1 to 13
25 mm
4 mm
316 Stainless Steel
10 μm
5000 psi
Hypersil™ HPLC Column
PRP-X100