missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Microlab™ Accessories - PTFE Tubing Assemblies

For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System

Brand:  Hamilton™ 240133

Code : NEW

Additional Details : Weight : 3.17510kg

 View more versions of this product

Product Code. 12698333

  • £35.40 / Each


Estimated Shipment
14-06-2018
Add to basket

Description

Description

  • For use with Hamilton Microlab 600 Diluters/Dispenser or Microlab 300 Pipetting System
Specifications

Specifications

Microlab™ 600 diluter/dispenser system
1372mm
PTFE Tubing Assemblies
1/4-28 Tubing threads
FEP
5.15mL
Videos