missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Microlab™ 600 Diluter/Dispenser System

Dilute/Dispense with better than 99% accuracy, independent of a liquid's viscosity, vapor pressure, and temperature. Microlab™ 600 Diluter/Dispenser System is a highly precise syringe pump with a graphical user interface designed to quickly and easily dilute and dispense volumes from 10μL to 50mL.

Brand:  Hamilton™ ML615-DIS


Additional Details : Weight : 8.00000kg

 View more versions of this product

Product Code. 16447820

  • £7080.00 / Each
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

 

  • Available as a single or dual syringe system
  • High torque, precision stepper motors deliver positional accuracy
  • Supports syringes from 10μL to 50mL
  • Ethernet port for communication with controller or corporate network
  • Printer Kit available

 

Syringe Pump Features

  • High torque valve motors
  • Precision syringe drives with 48,000 step resolution over 60 minutes
  • Lighted power and prime buttons
  • Independent left and right trigger ports
  • Fanless heat vent
  • 24V power input
  • CAN Daisy Chain input/out
  • RS-232 console port
  • Power over Ethernet (PoE)
  • TTL Input/Output

 

Dual Syringe Diluter

  • Creates up to a 1:50,000 dilution in single step
  • Reduces preparation time and wasted buffer
  • Washes tubing between each sample
Dispenser/Diluter Printer Kit
  • Prints on standard, non-thermal 8 1/2 × 11 and A4 paper
  • Interfaces with the free compliance software package that is available for Advanced Microlab™ devices
  • Satisfies regulatory CFR21 Part 11 and FDA GMP/GLP requirements by conveniently printing all log files directly from the device's controller

 

Dispenser: Sample spiking, Titration, Animal dosing (oral gavage), Reagent addition to chemical reactors, Flow chemistry, Manufacturing e.g. Mercury dispensing for light bulbs, Ink cartridge filling, Glue and epoxy dispensing, Nasal therapeutics (viscous), Contact lenses
Diluter: Forensics blood alcohol analysis and urinalysis, minig assay labs metals detection, environmental analysis water and soils testing, oil analysis preventative maintenance, alcohol testing beer and wine manufacturing

Specifications

Specifications

Basic controller
Dual Syringe Dispenser
Microlab 600
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers