Learn More
Hamilton™ Microlab 300 Pipetting System Accessories - Syringes
Choose from two syringes designed for use specifically with the Hamilton Microlab 300 Pipetting System
Brand: Hamilton™ 66088-15
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Model 1705.5
- Bubble Free Prime
- Expels air from the syringe and valve thanks to an innovative plunger tip
- 1.25mL
- Model 1002.5
- Gastight
- Ideal for dispensing both liquids and gases
- Has a precision-machined PTFE plunger tip which creates a leak-free seal
- 25μL
Specifications
1705.5 BFP | |
25 μL | |
Microlab™ 300 pipetteting system | |
Plunger tip |