Learn More
Hamilton™ Microlab 300 Pipetting System Accessories - Cable Management System
Keeps cables neat and safely tucked away
Brand: Hamilton™ 65164-01
Code : NEW
Additional Details : Weight : 0.20000kg
Description
- Complete system includes cable manger, probe holder, and accessory holder
- System components may be purchased separately
- For use with the Hamilton Microlab 300 Pipetting System
Specifications
Thumb Screw | |
Cable management system of Microlab™ 300 pipetteting system |