Learn More
Thermo Scientific™ Halt™ Protease Inhibitor Cocktail, EDTA-Free (100X)
Inhibit proteolytic degradation during cell lysis and protein extraction with these ready-to-use concentrated stock solutions of protease inhibitors.
Brand: Thermo Scientific™ 78437
208.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : PN
Additional Details : Weight : 0.80000kg
Description
Thermo Scientific™ Halt Protease Inhibitor Cocktails are ready-to-use concentrated stock solutions of protease inhibitors for addition to samples to prevent proteolytic degradation during cell lysis and protein extraction procedures.
Halt Protease Inhibitor Cocktails are 100X solutions containing optimized concentrations of six broad-spectrum protease inhibitors stabilized in high-quality dimethylsulfoxide (DMSO). The cocktails effectively inhibit serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The identity and concentration of each component is fully disclosed, leaving no uncertainties about variables and compatibility with experiments. Regular and EDTA-free formulations provide for inhibition of metalloproteases and compatibility with 2D electrophoresis, respectively. The stable, concentrated liquid cocktails are easy to use, and the several different package sizes make Halt Protease Inhibitor Cocktails exceptionally convenient for either small-scale or large-scale uses.Highlights:
Better than tablets – the refrigerator-stable, 100X liquid format is more effective and easier to use than individual inhibitors or other formats; just pipette exactly the amount you need
No proprietary ingredients – fully disclosed formulation contains AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A in DMSO
Multiple package sizes – choose among several package sizes, including single-use microtubes that keep protease inhibitors fresh and reduce the risk of reagent contamination
Two popular formulations – regular formulation includes a separate vial of EDTA solution for optional metalloprotease inhibition; EDTA-free formulation ensures compatibility with isoelectric focusing
Lysis buffer compatible – use with Thermo Scientific Pierce™ Cell Lysis Buffers or nearly any other commercial or homemade detergent-based lysis reagent
Includes:
Protease inhibitor cocktails with EDTA include separate 0.5M EDTA solution.
Specifications
Halt Protease Inhibitor Cocktail, EDTA-Free | |
Upon receipt store at 4°C | |
Halt™ | |
Protease Inhibitor | |
5 mL |
Protease Inhibitor Cocktail | |
Liquid | |
Lysis Buffer | |
Protease Inhibitor |
Safety and Handling
- Protease Inhibitor Cocktail
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 4-(2-aminoethyl)-benzenesulphonyl fluoride hydrochloride
For Research Use Only. Not for use in diagnostic procedures.