missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Haloenol Lactone Suicide Substrate, ≥98%
For cellular and molecular biology applications
POA - £246.00
Specifications
Color | White |
---|---|
Form | Lyophilized |
For Use With (Application) | Irreversibly inhibits calcium-independent phospholipase A-2 |
Molecular Weight (g/mol) | 317.18 |
Storage Requirements | -30° to -10°C |
Product Code | Brand | Substrate Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Substrate Quantity | Price | Quantity & Availability | |||||
15415919
|
Thermo Scientific™
J63728.MB |
25mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15405919
|
Thermo Scientific™
J63728.MA |
10mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15495909
|
Thermo Scientific™
J63728.#0 |
5mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
White | |
Irreversibly inhibits calcium-independent phospholipase A-2 | |
-30° to -10°C |
Lyophilized | |
317.18 |
For Research Use Only