missing translation for 'onlineSavingsMsg'
Learn More

HAI-1, clone: 9B10, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9960-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15288277

  • £43.63 / 25µg


Estimated Shipment
02-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The monoclonal antibody 9B10 recognizes HAI-1 also known as hepatocyte growth factor activation inhibitor. HAI-1 is a single transmembrane protein of approximately 53kDa with an extracellular Kunitz-type serine protease inhibitor domain. HAI-1 and HAI-2 are produced in membrane-associated forms, which are secreted as active, proteolytically truncated proteins. HAI can bind to and inhibit HGFA and matripase, which are responsible for converting HGF to an active form. Human HAI-1 transcript is expressed in various human tissues, such as adult placenta, kidney, pancreas, prostate, small intestine, fetal kidney and fetal lung. HAI-1 is a marker for prostate and breast cancers.Applications Reported: This 9B10 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunoblotting (WB) under nonreducing conditions only.Applications Tested: This 9B10 antibody has been tested by flow cytometric analysis of human cell line MCF7. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

HAI-1
9B10
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
HGF activator inhibitor-1, hepatocyte growth factor
IgG2a
RUO
Primary
Human
Videos