missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) oxide, tech., Thermo Scientific Chemicals

Catalog Number p-7051014
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
12055-23-1
HfO2
210.49
MFCD00003565
WIHZLLGSGQNAGK-UHFFFAOYSA-N
hafnium(4+) dioxidandiide
[O--].[O--].[Hf+4]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
12055-23-1
HfO2
210.49
MFCD00003565
WIHZLLGSGQNAGK-UHFFFAOYSA-N
hafnium(4+) dioxidandiide
[O--].[O--].[Hf+4]

Hafnium(IV) oxide is used in optical coatings and in advanced metal-oxide-semiconductor devices. It is used as a gate insulator in field-effect transistors due to its high dielectric constant. It plays an important role as a possible candidate for resistive-switching memories, as a refractory material in the insulation of devices such as thermocouples and as a high-κ dielectric in DRAM capacitors. It is also used in the preparation of hafnium tetrachloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 2774°C
Physical Form Powder
Quantity 100 g
Merck Index 14,4588
Solubility Information Insoluble in water.
Formula Weight 210.49
Grade Technical
Odor Odorless
Density 9.68 g/mL
Chemical Name or Material Hafnium(IV) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'