missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Hafnium iodide, ultra dry, 98.5% (metals basis)

Brand:  Alfa Aesar™ 035733.04

Code : NEW

Additional Details : CAS Number : 13777-23-6 Weight : 0.00200kg Transport : UN number : 3260 Chem class : 1a Pack group : II

Quantity 2g

 View more versions of this product

Product Code. 11435050

  • £81.30 / 2g


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

Hafnium iodide
Ultra dry
MFCD00016128
13777-23-6
686.11
UN3260

Specifications

Hafnium iodide
13777-23-6
98.5%
686.11
UN3260
Moisture Sensitive
2g
Ultra dry
Powder
(metals basis)
MFCD00016128
Ampule
400°C (vac.) (subl.)
Videos
SDS