missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Hafnium iodide, ultra dry, 98.5% (metals basis)

Brand:  Alfa Aesar™ 035733.09

UNSPSC : 12141718

Code : NEW

Additional Details : CAS Number : 13777-23-6 Weight : 0.01000kg Transport : UN number : 3260 Chem class : 1a Pack group : II

Quantity 10g

 View more versions of this product

Product Code. 11445050

  • £365.70 / 10g


Estimated Shipment date
from Supplier 25-08-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

Hafnium iodide
Ultra dry
MFCD00016128
13777-23-6
686.11
UN3260

Specifications

686.11
Ampule
Powder
UN3260
Hafnium iodide
Moisture Sensitive
13777-23-6
400°C (vac.) (subl.)
Ultra dry
98.5%
10g
(metals basis)
MFCD00016128
Documentation

Technical Documents