missing translation for 'onlineSavingsMsg'
Learn More

Hafnium foil, 0.025mm (0.001in) thick, 99.5% (metals basis excluding Zr), Zr nominal 3%, Thermo Scientific Chemicals

Catalog Number 11304677
Click to view available options
Quantity:
100 x 100 mm
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-58-6
Hf
178.49
MFCD00011032
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
23986
CHEBI:33343
hafnium
[Hf]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7440-58-6
Hf
178.49
MFCD00011032
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
23986
CHEBI:33343
hafnium
[Hf]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 100 x 100 mm
Percent Purity 99.5%
Odor Odorless
Weight ≈3.3g/100x100mm
Form Foil
Assay (metals basis excluding Zr), Zr nominal 3%
Chemical Name or Material Hafnium foil, 0.025mm (0.001 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'