missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hafnium ethoxide, 99.9% (metals basis), Thermo Scientific Chemicals
102.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
Hafnium ethoxide is a chemical used in the preparation of refractory coatings. Some of the other applications include Nano-materials & Thin Films, Display & Imaging, Catalysis & Synthesis, Hybrid & Disperse Materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 178°C to 180°C |
Boiling Point | 180°C to 200°C (13 mmHg) |
Quantity | 5 g |
UN Number | UN3180 |
Sensitivity | Moisture sensitive |
Solubility Information | It is soluble in hydrocarbons,reacts with ketones,esters. Reacts exothermally with water |
Formula Weight | 358.73 |
Percent Purity | 99.9% |
Physical Form | Crystalline |
Assay | (metals basis) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'