missing translation for 'onlineSavingsMsg'
Learn More

Hafnium arc-cast pellet, 12mm (0.47 in.) dia., 99.8% (metals basis excluding Zr), Zr nominal 4.5%, Thermo Scientific™

Catalog Number 15414945
Click to view available options
Quantity:
1 Pc.
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 7440-58-6
Melting Point 2227°C
Boiling Point 4603°C
Density 13.310 g/mL
Molecular Formula Hf
MDL Number MFCD00011032
Quantity 1 Pc.
Synonym hafnium, elemental, hafnio, celtium, unii-x71938l1do, and compounds, hsdb 552, powder, foil, atom, wire
InChI Key VBJZVLUMGGDVMO-UHFFFAOYSA-N
SMILES [Hf]
IUPAC Name hafnium
Molecular Weight (g/mol) 178.49
PubChem CID 23986
ChEBI CHEBI:33343
Formula Weight 178.49
Percent Purity 99.8%
Odor Odorless
Weight ≈10g/pellet
Form Pellet
Assay (metals basis excluding Zr), Zr nominal 4.5%
Chemical Name or Material Hafnium arc-cast pellet, 12mm (0.47 in.) dia.
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'