missing translation for 'onlineSavingsMsg'
Learn More

HA Recombinant Influenza A Virus Protein, subtype H1N1, (A/California/04/2009), His Tag

HA Recombinant Influenza A Virus Protein, subtype H1N1, (A/California/04/2009), His Tag

Brand:  Invitrogen 11055V08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13436066

  • £666.00 / 250µg

Estimated Shipment
02-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Hemagglutinin, Influenza A virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 529) of influenza A virus, subtype H1N1, (A/California/04/2009), HA (ACP41105.1) fused to a polyhistidine tag at the C-terminus. Subunits HA1 and HA2 remain uncleaved.

Activity: This recombinant protein has not been tested.

Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein assays and analysis
Lyophilized
HA Recombinant Influenza A Virus Subtype H1N1 (A/California/04/2009) Protein
5 x 50μg
For research use only. Not for use in diagnostic procedures
Viral proteins
HA Recombinant Influenza A Virus Subtype H1N1 (A/California/04/2009) Protein
Assay standard
Mammalian
Influenza
Lyophilized
59kDa
≥85% by SDS-PAGE
–20 to –70°C
5 Pack(s)
HA
HA
Other proteins
HA Recombinant Influenza A Virus Subtype H1N1 (A/California/04/2009) Protein
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.