Learn More
TCR H-Y (male antigen) Monoclonal Antibody (T3.70), FITC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£84.00 - £174.00
Specifications
Antigen | TCR H-Y (male antigen) |
---|---|
Clone | T3.70 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15248389
|
Affymetrix eBioscience
11-9930-82 |
100 μg |
£174.00
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15238389
|
Affymetrix eBioscience
11-9930-81 |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The T3.70 monoclonal antibody reacts with the transgenic alpha betaTCR that recognizes the male antigen H-Y in the context of H-2D^b. A large fraction of T cells in H-Y TCR transgenic mouse expresses this receptor. The H-Y TCR transgenic mouse has been used extensively to study T cell development and the role of thymic major histocompatibility complex in CD4^+ and CD8^+ T cell differentiation. Applications Reported: The T3.70 antibody has been reported for use in flow cytometric analysis. Applications Tested: The T3.70 antibody has been tested by flow cytometric analysis of CD11.3 line, which is derived from a thymic tumor of the H-Y TCR transgenic mice and expresses the transgenic H-Y TCR. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
A large fraction of T cells in H-Y TCR transgenic mouse expresses this receptor. The H-Y TCR transgenic mouse has been used extensively to study T cell development and the role of thymic major histocompatibility complex in CD4+ and CD8+ T cell differentiation.Specifications
TCR H-Y (male antigen) | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Male antigen HY; TCR HY | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
T3.70 | |
Flow Cytometry | |
FITC | |
Mouse | |
Mouse | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.