Learn More
H-9 dihydrochloride, Tocris Bioscience™
Protein kinase inhibitor
Brand:  Tocris (Bio-Techne) 039610
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487613
  • £114.75 / 10mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
Protein kinase inhibitor. Inhibits PKA (Ki = 1.9 μM), PKG (Ki = 0.9 μM), CaMK II (Ki = 60 μM), PKC (Ki = 18 μM), casein kinase I (Ki = 110 μ M) and casein kinase II (Ki > 300 μM).
Specifications
Chemical Identifiers
116700-36-8
324.22
N-(2-Aminoethyl)-5-isoquinolinesulfonamide dihydrochloride
N-(2-aminoethyl)isoquinoline-5-sulfonamide;dihydrochloride
C11H15Cl2N3O2S
HBLCYSFLYMHCBM-UHFFFAOYSA-N
11957465
C1=CC2=C(C=CN=C2)C(=C1)S(=O)(=O)NCCN.Cl.Cl
Specifications
H-9 dihydrochloride
>99%
C11H15Cl2N3O2S
HBLCYSFLYMHCBM-UHFFFAOYSA-N
N-(2-aminoethyl)isoquinoline-5-sulfonamide;dihydrochloride
11957465
116700-36-8
10mg
N-(2-Aminoethyl)-5-isoquinolinesulfonamide dihydrochloride
C1=CC2=C(C=CN=C2)C(=C1)S(=O)(=O)NCCN.Cl.Cl
324.22
324.22
Videos