Learn More
Tocris Bioscience™ H-7 dihydrochloride
Protein kinase inhibitor
Brand:  Tocris Bioscience™ 054210
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13427703
  • £117.60 / 10mg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Protein kinase inhibitor. IC50 values for inhibition of PKC, PKG, PKA and myosin light chain kinase are 6.0, 5.8, 3.0 and 97.0 μM respectively.
Specifications
Specifications
'&plusmin;-1-(5-Isoquinolinesulphonyl)-2-methylpiperazine dihydrochloride
>99%
Observed MW: 364.29
Soluble to 20mM in DMSO
108930-17-2
C14H17N3O2S.2HCl