missing translation for 'onlineSavingsMsg'
Learn More

Edulab Gyroscope for showing the effect of precession brass 75mm diameter, 30mm rim

Brand:  Edulab PH0342

Gyroscope for showing the effect of precession brass 75mm diameter, 30mm rim

Code : 50

Additional Details : Weight : 1.00000kg

Product Code. 13192760

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.