Learn More
GW 9662, Tocris Bioscience™
Selective covalent PPARγ antagonist
Brand:  Tocris (Bio-Techne) 150810
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13469573
  • £155.82 / 10mg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
Selective irreversible PPARγ antagonist (IC50 values are 3.3, 32 and 2000 nM for PPARγ, PPARα and PPARδ respectively). Blocks the inhibition of osteoclast formation induced by IL-4 in the low micromolar range (1-2 μM), therefore is more potent than BADGE (Cat. No. 1326). Anticancer, inhibits growth of human mammary tumor cell lines.
Specifications
Chemical Identifiers
22978-25-2
276.676
2-chloro-5-nitrobenzanilide, 2-chloro-5-nitro-n-phenyl-benzamide, 2-chloro-5-nitro-n-4-phenylbenzamide, 2-chloro-5-nitrophenyl-n-benzamide, tocris-1508, spectrum5_001989, lopac-m-6191, d00acx, d05thb, dsstox_cid_20723
CHEBI:79993
C1=CC=C(C=C1)NC(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl
C13H9ClN2O3
DNTSIBUQMRRYIU-UHFFFAOYSA-N
644213
2-chloro-5-nitro-N-phenylbenzamide
Specifications
10mg
22978-25-2
2-chloro-5-nitrobenzanilide, 2-chloro-5-nitro-n-phenyl-benzamide, 2-chloro-5-nitro-n-4-phenylbenzamide, 2-chloro-5-nitrophenyl-n-benzamide, tocris-1508, spectrum5_001989, lopac-m-6191, d00acx, d05thb, dsstox_cid_20723
C1=CC=C(C=C1)NC(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl
276.676
CHEBI:79993
>98%
GW 9662
C13H9ClN2O3
DNTSIBUQMRRYIU-UHFFFAOYSA-N
2-chloro-5-nitro-N-phenylbenzamide
644213
276.68