missing translation for 'onlineSavingsMsg'
Learn More

GW 791343 hydrochloride, Tocris Bioscience™

P2X7 allosteric modulator

Brand:  Tocris (Bio-Techne) 338550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13442924

  • £546.02 / 50mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

P2X7 allosteric modulator. Exhibits species-specific activity; acts as a negative allosteric modulator of human P2X7 (pIC50 = 6.9 - 7.2) and a positive allosteric modulator of rat P2X7.
Specifications

Chemical Identifiers

309712-55-8
483.81
gw 791343 hydrochloride, gw791343 trihydrochloride, gw791343 trihydrochloride, 2-3,4-difluorophenyl amino-n-2-methyl-5-1-piperazinylmethyl phenyl-acetamide trihydrochloride, 2-3,4-difluorophenyl amino-n-2-methyl-5-piperazin-1-ylmethyl phenyl acetamide trihydrochloride, d05exn, gw-791343 hydrochloride, gw 791343 trihydrochloride, c20h24f2n4o.3clh, gw 791343 trihydrochloride;gw-791343 trihydrochloride
2-(3,4-difluoroanilino)-N-[2-methyl-5-(piperazin-1-ylmethyl)phenyl]acetamide;trihydrochloride
C20H27Cl3F2N4O
WSBRAHWNJBXXJM-UHFFFAOYSA-N
9848159
CC1=C(C=C(C=C1)CN2CCNCC2)NC(=O)CNC3=CC(=C(C=C3)F)F.Cl.Cl.Cl

Specifications

GW 791343 hydrochloride
309712-55-8
gw 791343 hydrochloride, gw791343 trihydrochloride, gw791343 trihydrochloride, 2-3,4-difluorophenyl amino-n-2-methyl-5-1-piperazinylmethyl phenyl-acetamide trihydrochloride, 2-3,4-difluorophenyl amino-n-2-methyl-5-piperazin-1-ylmethyl phenyl acetamide trihydrochloride, d05exn, gw-791343 hydrochloride, gw 791343 trihydrochloride, c20h24f2n4o.3clh, gw 791343 trihydrochloride;gw-791343 trihydrochloride
CC1=C(C=C(C=C1)CN2CCNCC2)NC(=O)CNC3=CC(=C(C=C3)F)F.Cl.Cl.Cl
483.81
483.81
50mg
C20H27Cl3F2N4O
WSBRAHWNJBXXJM-UHFFFAOYSA-N
2-(3,4-difluoroanilino)-N-[2-methyl-5-(piperazin-1-ylmethyl)phenyl]acetamide;trihydrochloride
9848159
>99%
Videos