missing translation for 'onlineSavingsMsg'
Learn More

GW 7647, Tocris Bioscience™

Highly selective, potent PPARα agonist. Orally active

Brand:  Tocris (Bio-Techne) 167710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13459903

  • £251.37 / 10mg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and highly selective PPARα agonist (EC50 values are 6, 1100 and 6200 nM for human PPARα, PPARγ and PPARδ receptors respectively). Modulates oleate metabolism and mitochondrial enzyme gene expression in mature myotubules in vitro. Has lipid-lowering effects following oral administration in vivo. Reduces NO production in macrophages; exhibits anti-inflammatory properties.
Specifications

Chemical Identifiers

265129-71-3
502.758
chembl21241, 2-4-2-4-cyclohexylbutyl cyclohexylcarbamoyl amino ethyl phenyl sulfanyl-2-methylpropanoic acid, 2-4-2-1-cyclohexanebutyl-3-cyclohexylureido ethyl-phenyl-thio-2-methyl-propionic acid, 2-4-2-cyclohexylamino carbonyl 4-cyclohexylbutyl amino ethyl phenyl thio-2-methylpropanoic acid, 2-4-2-1-cyclohexylbutyl-3-cyclohexylureido ethyl phenylthio-2-methylpropionic acid, 2-4-2-1-4-cyclohexylbutyl-3-cyclohexylureido ethyl phenylthio-2-methylpropionic acid, 2-4-2-1-cyclohexanebutyl-3-cyclohexylureido ethyl phenylthio-2-methylpropionic acid, tocris-1677
CHEBI:70778
CC(C)(C(=O)O)SC1=CC=C(C=C1)CCN(CCCCC2CCCCC2)C(=O)NC3CCCCC3
C29H46N2O3S
PKNYXWMTHFMHKD-UHFFFAOYSA-N
3392731
2-[4-[2-[4-cyclohexylbutyl(cyclohexylcarbamoyl)amino]ethyl]phenyl]sulfanyl-2-methylpropanoic acid

Specifications

10mg
265129-71-3
chembl21241, 2-4-2-4-cyclohexylbutyl cyclohexylcarbamoyl amino ethyl phenyl sulfanyl-2-methylpropanoic acid, 2-4-2-1-cyclohexanebutyl-3-cyclohexylureido ethyl-phenyl-thio-2-methyl-propionic acid, 2-4-2-cyclohexylamino carbonyl 4-cyclohexylbutyl amino ethyl phenyl thio-2-methylpropanoic acid, 2-4-2-1-cyclohexylbutyl-3-cyclohexylureido ethyl phenylthio-2-methylpropionic acid, 2-4-2-1-4-cyclohexylbutyl-3-cyclohexylureido ethyl phenylthio-2-methylpropionic acid, 2-4-2-1-cyclohexanebutyl-3-cyclohexylureido ethyl phenylthio-2-methylpropionic acid, tocris-1677
CC(C)(C(=O)O)SC1=CC=C(C=C1)CCN(CCCCC2CCCCC2)C(=O)NC3CCCCC3
502.758
CHEBI:70778
>99%
GW 7647
C29H46N2O3S
PKNYXWMTHFMHKD-UHFFFAOYSA-N
2-[4-[2-[4-cyclohexylbutyl(cyclohexylcarbamoyl)amino]ethyl]phenyl]sulfanyl-2-methylpropanoic acid
3392731
502.75
Videos
Documents