Learn More
GW 542573X, Tocris Bioscience™
Selective activator of KCa2.1 channels
Brand:  RD Systems (Bio-Techne) 4311/50
Code : Z1
Additional Details : Weight : 0.00050kg
Quantity 50mg
 View more versions of this product
Product Code. 13454544
  • £787.92 / 50mg
Estimated Shipment date
from Supplier 24-03-2020
Description
Description
Activator of small-conductance Ca2+-activated K+ channels (KCa2); selective for KCa2.1 (EC50 = 8.2 μM in HEK293 cells expressing hKCa2.1) with the profile hKCa2.1 > hKCa2.2 = hKCa2.3 > hKCa3.1 (IK).
Specifications
Chemical Identifiers
660846-41-3
364.442
tert-butyl 4-2-methoxyphenyl carbamoyl oxy methyl piperidine-1-carboxylate, gw542573x hplc, 4-2-methoxyphenylcarbamoyloxymethyl-piperidine-1-carboxylic acid t-butyl ester, 4-2-methoxyphenyl amino carbonyl oxy methyl-piperidinecarboxylic acid-1,1-dimethylethyl ester
tert-butyl 4-[(2-methoxyphenyl)carbamoyloxymethyl]piperidine-1-carboxylate
C19H28N2O5
SAXGSDIZIYFNKD-UHFFFAOYSA-N
24885053
CC(C)(C)OC(=O)N1CCC(CC1)COC(=O)NC2=CC=CC=C2OC
Specifications
50mg
660846-41-3
tert-butyl 4-2-methoxyphenyl carbamoyl oxy methyl piperidine-1-carboxylate, gw542573x hplc, 4-2-methoxyphenylcarbamoyloxymethyl-piperidine-1-carboxylic acid t-butyl ester, 4-2-methoxyphenyl amino carbonyl oxy methyl-piperidinecarboxylic acid-1,1-dimethylethyl ester
CC(C)(C)OC(=O)N1CCC(CC1)COC(=O)NC2=CC=CC=C2OC
364.442
364.44
GW 542573X
C19H28N2O5
SAXGSDIZIYFNKD-UHFFFAOYSA-N
tert-butyl 4-[(2-methoxyphenyl)carbamoyloxymethyl]piperidine-1-carboxylate
24885053
>99%