missing translation for 'onlineSavingsMsg'
Learn More

GW 1929 hydrochloride, Tocris Bioscience™

Selective PPARγ agonist. Orally active

Brand:  Tocris (Bio-Techne) 166450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13439883

  • £705.24 / 50mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Highly selective orally active peroxisome proliferator-activated receptor (PPAR)γ agonist (pEC50 values are 8.05, < 4 and < 4 for human PPARγ, PPARα and PPARδ receptors respectively). Decreases glucose, fatty acid and triglyceride levels following oral administration in vivo.
Specifications

Chemical Identifiers

1217466-21-1
532.037
gw 1929 hydrochloride, n-2-benzoylphenyl-o-2-methyl-2-pyridinylamino ethyl-l-tyrosine hydrochloride, 2s-2-2-benzoylphenyl amino-3-4-2-methyl pyridin-2-yl amino ethoxy phenyl propanoic acid hydrochloride
(2S)-2-(2-benzoylanilino)-3-[4-[2-[methyl(pyridin-2-yl)amino]ethoxy]phenyl]propanoic acid;hydrochloride
C30H30ClN3O4
KXNKIKXTGRMLEY-YCBFMBTMSA-N
56972174
CN(CCOC1=CC=C(C=C1)CC(C(=O)O)NC2=CC=CC=C2C(=O)C3=CC=CC=C3)C4=CC=CC=N4.Cl

Specifications

GW 1929 hydrochloride
C30H30ClN3O4
KXNKIKXTGRMLEY-YCBFMBTMSA-N
(2S)-2-(2-benzoylanilino)-3-[4-[2-[methyl(pyridin-2-yl)amino]ethoxy]phenyl]propanoic acid;hydrochloride
56972174
>98%
1217466-21-1
gw 1929 hydrochloride, n-2-benzoylphenyl-o-2-methyl-2-pyridinylamino ethyl-l-tyrosine hydrochloride, 2s-2-2-benzoylphenyl amino-3-4-2-methyl pyridin-2-yl amino ethoxy phenyl propanoic acid hydrochloride
CN(CCOC1=CC=C(C=C1)CC(C(=O)O)NC2=CC=CC=C2C(=O)C3=CC=CC=C3)C4=CC=CC=N4.Cl
532.037
532.03
50mg
Videos