missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Guko Rubber Gasket

Robu™ Guko Rubber Gasket is a conical gasket suitable for use with filter funnels.

Brand:  Robu™ 21856

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10032211

  • £3.70 / Each


Estimated Shipment
09-07-2020
Add to basket

Description

Description

  • Guko
  • Single
  • Conical gasket
  • Suitable for filter funnels
Specifications

Specifications

35 mm
Single Gasket
64 mm
Conical
Rubber
48 mm outer, 32 mm inner
Funnel
Gasket
Videos
Documents