Learn More
GUCA2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553757
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SLESVKKLKD LQEPQEPRVG KLRNFAPIPG EPVVPILCSN PNFPEELKPL CKEPNAQEIL QRLEEIAEDP GTCEICAYAA CTGC Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 64%.
GUCA2A Polyclonal antibody specifically detects GUCA2A in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
GUCA2A | |
Polyclonal | |
Unconjugated | |
GUCA2A | |
GCAP-I; GUANYL; guanylate cyclase activator 2 (guanylin 2, intestinal, heatstable); guanylate cyclase activator 2 (guanylin); guanylate cyclase activator 2A; guanylate cyclase activator 2A (guanylin 2, intestinal, heat-stable); guanylate cyclase activator 2a (guanylin); guanylate cyclase-activating protein 1; Guanylate cyclase-activating protein I; guanylin; GUCA2; Guca2a; HMW-guanylin; prepro-guanylin; STARA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2980 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q02747 | |
GUCA2A | |
Recombinant protein corresponding to Human GUCA2A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only