missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanine hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Guanine hydrochloride is one of the strongest denaturant. It is mainly used in protein purification and protein folding studies. It is a nucleobase used in biochemical research.
Solubility
Slightly soluble in water
Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents.
Guanine hydrochloride is one of the strongest denaturant. It is mainly used in protein purification and protein folding studies. It is a nucleobase used in biochemical research.
Solubility
Slightly soluble in water
Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Melting Point | >300°C |
Assay Percent Range | 99% |
Quantity | 10 g |
Beilstein | 4159901 |
Merck Index | 14,4564 |
Solubility Information | Slightly soluble in water |
Formula Weight | 187.59 |
Percent Purity | 99% |
Chemical Name or Material | Guanine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'