Learn More
Guanidine Hydrochloride (Colorless-to-White Crystals), Fisher BioReagents™
For isolation of RNA from cell extracts
Brand: Fisher Bioreagents BP178-1
137.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 32
Additional Details : CAS Number : 50-01-1 Weight : 1.05000kg
Description
For isolation of RNA from cell extracts
Guanidine Hydrochloride is a strong protein-denaturing agent used in the isolation of RNA from cell extracts. RNase activity is inhibited in the presence of this reagent.Specifications
Guanidine Hydrochloride | |
≥99.5% | |
1 kg | |
178°C | |
95.53 | |
White |
50-01-1 | |
guanidine hydrochloride | |
15, 4597 | |
CH6ClN3 | |
Solid | |
≥99.5% |
Safety and Handling
- Guanidine hydrochloride, without anticaking agent
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.