missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Carbonate, 99+%, Thermo Scientific Chemicals

Catalog Number 13445748
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
593-85-1
0·5 H2CO3
90.08
MFCD00013029
This item is not returnable. View return policy
593-85-1
0·5 H2CO3
90.08
MFCD00013029

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 267.0°C to 270.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic drum
Linear Formula [C(NH2)3]2CO3
Quantity 5 kg
Beilstein 03,82
Solubility Information Solubility in water: 450g/L (20 c). Other solubilities: slightly soluble in alcohol and acetone
Formula Weight 90.08
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Guanidine carbonate, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'