missing translation for 'onlineSavingsMsg'
Learn More

Guaiazulene, 99%, ACROS Organics™

Brand:  Acros Organics 120200250

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 489-84-9 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 11967031

  • £41.00 / 25g


Estimated Shipment
13-09-2017
Add to basket


Specifications

Chemical Identifiers

Guaiazulene
99%
198.31
99%
C15H18
1, 4-Dimethyl-7-isopropylazulene

Specifications

Guaiazulene
Authentic
99%
Glass bottle
198.31
1, 4-Dimethyl-7-isopropylazulene
Dark blue
153.0°C (7.0 Torr)
>110°C
489-84-9
Low Melting Crystalline Solid
99%
C15H18
Solubility in water: 25%. Other solubilities: soluble in ethanol, ethyl ether and ethyl acetate
0.9700g/mL
29.0° to 31.0°C
25g
Safety and Handling

Safety and Handling

Product Identifier
  • Guaiazulene

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documentation

Technical Documents