missing translation for 'onlineSavingsMsg'
Learn More

Guaiazulene, 99%, Thermo Scientific Chemicals

Catalog Number p-4228708
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
489-84-9
C15H18
198.31
FWKQNCXZGNBPFD-UHFFFAOYSA-N
guaiazulene, 1,4-dimethyl-7-isopropylazulene, azulon, vetivazulen, azunol, 7-isopropyl-1,4-dimethylazulene, eucazulen, guajazulene, kessazulen, purazulen
3515
CHEBI:5550
1,4-dimethyl-7-propan-2-ylazulene
CC1=C2C=CC(=C2C=C(C=C1)C(C)C)C
This item is not returnable. View return policy
489-84-9
C15H18
198.31
FWKQNCXZGNBPFD-UHFFFAOYSA-N
guaiazulene, 1,4-dimethyl-7-isopropylazulene, azulon, vetivazulen, azunol, 7-isopropyl-1,4-dimethylazulene, eucazulen, guajazulene, kessazulen, purazulen
3515
CHEBI:5550
1,4-dimethyl-7-propan-2-ylazulene
CC1=C2C=CC(=C2C=C(C=C1)C(C)C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 29.0°C to 31.0°C
Color Blue
Density 0.9700g/mL
Boiling Point 153.0°C (7.0 Torr)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 05,II,473
Merck Index 15,459
Specific Gravity 0.97
Solubility Information Solubility in water: 25%. Other solubilities: soluble in ethanol, ethyl ether and ethyl acetate
Formula Weight 198.31
Percent Purity 99%
Physical Form Low Melting Crystalline Solid
Chemical Name or Material Guaiazulene
Show More Show Less
compliance-icons
Product Identifier
  • Guaiazulene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'