missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2H3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562808
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FSPICTTCET AFKISLPPVL KAKK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
GTF2H3 interacts with HIV-1 Tat as a component of the HIV-1 transcription pre-initiation complex, but is released from the elongation complex which includes P-TEFb. It synergizes with HIV-1 Tat to induce transcription elongation from the HIV-1 LTR promoter.Specifications
GTF2H3 | |
Polyclonal | |
Unconjugated | |
GTF2H3 | |
34kDa; 5033417D07Rik; Basic transcription factor 2 34 kDa subunit; BTF2; BTF2 p34; BTF2-p34; C730029A10; D5Ertd679e; General transcription factor IIH polypeptide 3; general transcription factor IIH subunit 3; general transcription factor IIH, polypeptide 3; general transcription factor IIH, polypeptide 3, 34kDa; Gtf2h3; P34; TFB4; TFIIH; TFIIH basal transcription factor complex p34 subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2967 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13889 | |
GTF2H3 | |
Recombinant protein corresponding to Human GTF2H3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only