Learn More
Fisherbrand™ GT4 Benchtop Centrifuge Packs
The Fisherbrand™ GT4 Benchtop Centrifuge is the premium performance high capacity model available in ventilated or rapid pull-down refrigerated versions
£7502.38 - £9821.76
Specifications
Type Benchtop Centrifuge
Frequency 50/60 Hz
Drive System Direct Brushless Induction
Rotor Included Yes
Safety Features ClickSeal biocontainment lid
View More Specs
Products 4
Product Code Brand Model Refrigerated Price Quantity & Availability  
Product Code Brand Model Refrigerated Price Quantity & Availability  
15828722
View Documents Promotion Details
Fisherbrand™
GT4 3L No
£7502.38
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-01-2024
to see stock.
15838722
View Documents Promotion Details
Fisherbrand™
GT4R 3L Yes
£7502.38
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-01-2024
to see stock.
15848722
View Documents Promotion Details
Fisherbrand™
GT4 4L No
£7502.38
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-09-2022
to see stock.
15858722
View Documents Promotion Details
Fisherbrand™
GT4R 4L Yes
£9821.76
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Description
Description
  • Premium performance and capacity benchtop centrifuge models available in 3 and 4 litre options
  • Specially engineered for high throughput applications
  • Outstanding capacity and versatility accommodating up to 40x50mL or 96x15mL conical tubes (4L version)
  • Ventilated and refrigerated versions also available, with rapid temperature pull-downs
  • Three litre package includes a TX-750 rotor for speeds up to 3488xg
  • Four litre package includes a high-mass TX-1000 rotor providing excellent vibration and imbalance protection up to 3739xg, with adapters for spinning down tubes, plates or a combination of the two
  • One-touch, easy-to-use operation with pre-saved protocols
  • Backlit display interface with multi-language options
  • Rapid push-button rotor exchange with the Auto-Lock™ rotor swap system
  • Corrosion resistant and lubricating rotor-cross coating
  • Fitted with ClickSeal™ biocontainment lids
  • Glove and wipe-clean, detergent-friendly
  • Two year warranty
Specifications
Specifications
Benchtop Centrifuge
Direct Brushless Induction
ClickSeal biocontainment lid
50/60 Hz
Yes