Learn More
Fisherbrand™ GT2 Benchtop Centrifuge Packs
The mid-sized Fisherbrand™ GT2 Benchtop Centrifuge provides speed and versatility, reaching 4238xg for fast and efficient separations
£6720.00 - £8920.00
Specifications
Type Benchtop centrifuge
Hertz 50/60Hz
Drive System Direct brushless induction drive
Rotor Included Yes
Safety Features ClickSeal biocontainment lid
View More Specs
Products 2
Product Code Brand Model Refrigerated Price Quantity & Availability  
Product Code Brand Model Refrigerated Price Quantity & Availability  
15808722 View Documents Fisherbrand™
GT2 No £6720.00
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
15818722 View Documents Fisherbrand™
GT2R Yes £8920.00
Each View alternative products
Save Up to 
»
In Stock 7
Description
Description
  • Mid-size benchtop centrifuge with speed (reaching 4238xg) and versatility but with minimal lab footprint
  • Ventilated and refrigerated versions available, with rapid temperature pull-downs
  • Includes a TX-400 rotor with adapters accommodating 36x15mL or 16x50mL conical tubes, ideal for cell culture applications
  • One-touch, easy-to-use operation with pre-saved protocols
  • Backlit display interface with multi-language options
  • Rapid push-button rotor exchange with the Auto-Lock™ rotor swap system
  • Corrosion resistant and lubricating rotor-cross coating
  • Fitted with ClickSeal™ biocontainment lids
  • Glove and wipe-clean, detergent-friendly
  • Two year warranty
Specifications
Specifications
Benchtop centrifuge
Direct brushless induction drive
ClickSeal biocontainment lid
50/60Hz
Yes