missing translation for 'onlineSavingsMsg'
Learn More

GT 2016, Tocris Bioscience™

Selective H3 antagonist

Brand:  Tocris (Bio-Techne) 241910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13461174

  • £135.00 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

High affinity H3 receptor antagonist (Ki = 43.8 nM). Displays selectivity against H1 and H2 receptors (IC50 >10μM). Increases the release of histamine in the cerebral cortex. Displays no activity at histamine methyltransferase in vitro at concentrations up to 3μM. Brain penetrant.
Specifications

Chemical Identifiers

152241-24-2
317.477
chembl14812, 5-cyclohexyl-1-4-3h-imidazol-4-yl piperidin-1-yl pentan-1-one, d02yxq, 5-cyclohexyl-1-4-1h-imidazol-5-yl-1-piperidinyl-1-pentanone
5-cyclohexyl-1-[4-(1H-imidazol-5-yl)piperidin-1-yl]pentan-1-one
C19H31N3O
YTCGNPGLMAECND-UHFFFAOYSA-N
9839975
C1CCC(CC1)CCCCC(=O)N2CCC(CC2)C3=CN=CN3

Specifications

GT 2016
152241-24-2
chembl14812, 5-cyclohexyl-1-4-3h-imidazol-4-yl piperidin-1-yl pentan-1-one, d02yxq, 5-cyclohexyl-1-4-1h-imidazol-5-yl-1-piperidinyl-1-pentanone
C1CCC(CC1)CCCCC(=O)N2CCC(CC2)C3=CN=CN3
317.477
317.47
10mg
C19H31N3O
YTCGNPGLMAECND-UHFFFAOYSA-N
5-cyclohexyl-1-[4-(1H-imidazol-5-yl)piperidin-1-yl]pentan-1-one
9839975
>98%
Videos