Learn More
GE Healthcare GSTrap™ FF Column
Brand:  GE Healthcare 17-5281-05
Code : A0
Additional Details : Weight : 1.04000kg
Product Code. 11340212
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
GSTrap FF columns are prepacked Glutathione Sepharose Fast Flow columns for fast, convenient, one-step purification of glutathione S-transferase (GST) tagged proteins. GST-tagged proteins can be purified directly from pretreated bacterial lysates using GSTrap™ FF. GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione. The purification process preserves protein antigenicity and function. If desired, cleavage of the protein from GST can be achieved using a site-specific protease whose recognition sequence is located immediately upstream from the multiple cloning site on the pGEX plasmids. GST-tagged proteins can be detected using colorimetric or immunological methods. The medium, Glutathione Sepharose™ 4 Fast Flow, is also available as lab packages and is an excellent choice for scale-up. The columns can be operated with a syringe, peristaltic pump, or liquid chromatography system such as ÅKTA™ design or FPLC™ System.
  • Fast and simple one-step purification of GST-tagged proteins.
  • Mild elution conditions preserving protein antigenicity and function.
  • Easy scale-up by connecting the columns in series.
  • Sepharose 4 Fast Flow provides good flow properties.
GSTrap HP columns are prepacked with Glutathione Sepharose High Performance medium and provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins. GSTrap HP 1mL and 5mL columns are prepacked with Glutathione Sepharose High Performance medium, which provides high-resolution purifications, thus a more concentrated sample. GST-tagged proteins are purified from pretreated bacterial lysates by affinity chromatography using immobilized glutathione, such as Glutathione Sepharose High Performance. GST-tagged proteins are bound to the affinity medium, and impurities are removed by washing with binding buffer GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione. The purification process preserves protein antigenicity and function. If desired, cleavage of the protein from GST can be achieved using a site-specific protease whose recognition sequence is located immediately upstream from the multiple cloning site on the pGEX plasmids. GST-tagged proteins can be detected using colorimetric or immunological methods.
  • High resolution purification of GST-tagged proteins.
  • Mild elution conditions preserving protein antigenicity and function.
  • Fast, one-step purification.
  • Easy to use with syringe, pump or a chromatography system.
Specifications
Specifications
1mL
Videos