Learn More
Cytiva GST™ Tagged Package
For use in protein purification applications
Brand: Cytiva 29-0588-04
Code : NEW
Additional Details : Weight : 0.10000kg
Description
The GST package is designed for easy purification of GST-tagged proteins. Contents: GSTrap 4B (1 x 1mL) and HiTrap Desalting (1 x 5mL)Specifications
GST package enables convenient purification of GST-tagged proteins. | |
4°C to 30°C | |
Complete packsize | |
Chromatography Media |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.