missing translation for 'onlineSavingsMsg'
Learn More

GSK 789472 hydrochloride, Tocris Bioscience™

D3 antagonist. Also D2 partial agonist

Brand:  Tocris (Bio-Techne) 394010

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13433854

  • £135.24 / 10mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective dopamine D3 receptor antagonist and D2 partial agonist (pEC50 values are <5.5 and 8.3 respectively). Selective against the D4 receptor in both agonist and antagonist assays. Brain penetrant.
Specifications

Chemical Identifiers

1257326-24-1
295.811
gsk 789472 hydrochloride, 1-phenyl-3-piperidin-2-ylmethyl imidazolidin-2-one hydrochloride, 4,5-dihydro-1-phenyl-3-2-piperidinyl methylimidazol-2 1h-one
1-phenyl-3-(piperidin-2-ylmethyl)imidazolidin-2-one;hydrochloride
C15H22ClN3O
BYJUVPWUMBVSTK-UHFFFAOYSA-N
57347570
C1CCNC(C1)CN2CCN(C2=O)C3=CC=CC=C3.Cl

Specifications

10mg
1257326-24-1
gsk 789472 hydrochloride, 1-phenyl-3-piperidin-2-ylmethyl imidazolidin-2-one hydrochloride, 4,5-dihydro-1-phenyl-3-2-piperidinyl methylimidazol-2 1h-one
C1CCNC(C1)CN2CCN(C2=O)C3=CC=CC=C3.Cl
295.811
295.81
GSK 789472 hydrochloride
C15H22ClN3O
BYJUVPWUMBVSTK-UHFFFAOYSA-N
1-phenyl-3-(piperidin-2-ylmethyl)imidazolidin-2-one;hydrochloride
57347570
>99%
Videos
Documents