Learn More
GSK 4716, Tocris Bioscience™
Selective ERRγ and ERRβ agonist
Brand:  Tocris (Bio-Techne) 307550
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13462344
  • £368.97 / 50mg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Selective agonist at estrogen-related receptors ERRβ and ERRγ. Displays selectivity over ERRα and the classical estrogen receptors.
Specifications
Chemical Identifiers
101574-65-6
282.343
e-4-hydroxy-n'-4-isopropylbenzylidene benzohydrazide, 4-hydroxy-benzoic acid 4-isopropyl-benzylidene-hydrazide, 4-hydroxy-2-1e-4-1-methylethyl phenyl methylene hydrazide, 2-4-1-methylethyl phenyl methylene hydrazide 4-hydroxybenzoic acid, 2gpp, d0dx8t, gsk hplc, 4-hydroxy-n-e-4-isopropylphenyl methyleneamino benzamide, 4-hydroxy-n-e-4-propan-2-ylphenyl methylideneamino benzamide, 4-hydroxy-n-1e-4-propan-2-yl phenyl methylidene benzohydrazide
CHEBI:80000
CC(C)C1=CC=C(C=C1)C=NNC(=O)C2=CC=C(C=C2)O
C17H18N2O2
IKPPIUNQWSRCOZ-WOJGMQOQSA-N
5331325
4-hydroxy-N-[(E)-(4-propan-2-ylphenyl)methylideneamino]benzamide
Specifications
50mg
101574-65-6
e-4-hydroxy-n'-4-isopropylbenzylidene benzohydrazide, 4-hydroxy-benzoic acid 4-isopropyl-benzylidene-hydrazide, 4-hydroxy-2-1e-4-1-methylethyl phenyl methylene hydrazide, 2-4-1-methylethyl phenyl methylene hydrazide 4-hydroxybenzoic acid, 2gpp, d0dx8t, gsk hplc, 4-hydroxy-n-e-4-isopropylphenyl methyleneamino benzamide, 4-hydroxy-n-e-4-propan-2-ylphenyl methylideneamino benzamide, 4-hydroxy-n-1e-4-propan-2-yl phenyl methylidene benzohydrazide
CC(C)C1=CC=C(C=C1)C=NNC(=O)C2=CC=C(C=C2)O
282.343
CHEBI:80000
>99%
GSK 4716
C17H18N2O2
IKPPIUNQWSRCOZ-WOJGMQOQSA-N
4-hydroxy-N-[(E)-(4-propan-2-ylphenyl)methylideneamino]benzamide
5331325
282.34