Learn More
Tocris Bioscience™ GSK 429286
Selective Rho-kinase (ROCK) inhibitor
Brand:  Tocris Bioscience™ 372610
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13463494
  • £237.61 / 10mg
Estimated Shipment
03-04-2019
Add to basket
Description
Description
Selective Rho-kinase inhibitor (IC 50 values are 14, 780 and 1940 nM for ROCK1, RSK and p70S6K respectively). Reverses adrenalin-induced contraction of the rat aortic ring (IC 50 = 190 nM) and causes a dose-dependent decrease in mean arterial blood pressure in spontaneous hypertensive rats. Orally active.
Specifications
Specifications
GSK 429286
>97
10mg
GSK 429286
432.37