missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human GSK-3B (GSK3 beta), His Tag Recombinant Protein
Human GSK-3B (GSK3 beta), His Tag Recombinant Protein
Brand: Thermo Scientific™ PV3365
Code : 50
Additional Details : Weight : 0.05250kg
Description
GSK3B is a proline-directed serine⁄threonine protein kinase involved in energy metabolism, neuronal cell development and body pattern formation.Specifications
NP_002084 | |
2932 | |
10 μg | |
GSK Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CMGC Group | |
Z'-LYTE™, Omnia™ Bulk Substrate | |
glycogen synthase kinase 3 beta | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
53.1 kDa | |
GSK-3B (GSK3 beta) | |
Human | |
His Tag | |
Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
GSK3B | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |