missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSG2 (Haspin) Recombinant Human Protein
GSG2 phosphorylates histone H3 and is viewed as a potential therapeutic target for oncology.
Brand: Invitrogen™ PV5709
Code : NEW
Additional Details : Weight : 0.01000kg
Description
GSG2 (haspin) is a conserved kinase required for mitosis.
Specifications
NP_114171.1 | |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol. | |
1mg | |
RUO - research use only | |
GSG2 (Haspin) | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
64.8kDa | |
Kinase proteins are shipped on dry ice. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
Human | |
GST Tag | |
Frozen |