missing translation for 'onlineSavingsMsg'
Learn More

Rat Growth Hormone Antibody, R&D Systems™

Mouse Monoclonal Antibody

POA

Specifications

Antigen Growth Hormone
Clone 222517
Concentration LYOPH
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Applications Western Blot, Immunohistochemistry
View More Specs

Description

Description

Growth Hormone Monoclonal specifically detects Growth Hormone in Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Growth Hormone
LYOPH
Western Blot, Immunohistochemistry
Unconjugated
Mouse
Rat
P01244
2688
E. coli-derived recombinant rat Growth Hormone Phe27-Phe216 Accession # P01244
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects rat Growth Hormone in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Growth Hormone is observed.
222517
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
GH1, GHB5, GHIGHD1B, GHN, GH-N, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N, IGHD1A, IGHD2, Somatotropin
GH1
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only