missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRK7 Recombinant Human Protein

GRK7 is a serine/threonine kinase that is specifically expressed in the retina.

£1918.00 - £9080.00

Specifications

Format Frozen
Host Species Human
Protein Tag GST Tag
Recombinant Yes
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15458523 View Documents Invitrogen™
PR7131A
100μg £1918.00
100µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 19-06-2020
Please to purchase this item. Need a web account? Register with us today!
10221163 View Documents Invitrogen™
PV4125
1mg £9080.00
1mg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 19-06-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GRK7 phosphorylates cone opsins and initiates their deactivation.

Specifications

Specifications

Frozen
Human
GST Tag
Yes