missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRK7 Recombinant Human Protein
GRK7 is a serine/threonine kinase that is specifically expressed in the retina.
Brand: Invitrogen™ PR7131A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
GRK7 phosphorylates cone opsins and initiates their deactivation.
Specifications
NP_631948 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
GRK7, GPRK7 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
89.7kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
GRK Kinase Family | |
Recombinant | |
Baculovirus |