missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK5 Recombinant Human Protein
GRK5 Recombinant Human Protein
Brand: Invitrogen PV3824
Code : 50
Additional Details : Weight : 0.05250kg
Description
GRK5 is a serine⁄threonine kinase that regulates the activity of a variety of G protein-coupled receptors and the motility of polymorphonuclear leukocytes.Specifications
NP_005299 | |
Cell Viability, Pharma and Biopharma, Cell Proliferation and Function, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
95.2kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
GRK5, GPRK5 | |
GRK5 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 10μM ATP, 145μg/mL PLKtide peptide substrate (EVIEASFAEQEAK) and trace [32P]- γ-ATP for 10 minutes at 30°C | |
GRK5 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.26mg/mL | |
2869, 6775 | |
GRK5 Recombinant Human Protein | |
–80°C | |
1 Each | |
GRK5 Recombinant Human Protein | |
GRK | |
0.8nmoles of phosphate transferred to PLKtide peptide substrate (EVIEASFAEQEAK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 16.7μg/mL | |
GST Tag | |
Kinase and inhibitors | |
>85% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.