missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK4 Recombinant Human Protein
GRK4 Recombinant Human Protein
Brand: Invitrogen PV3807
Code : 50
Additional Details : Weight : 0.05250kg
Description
GRK4 is a member of the G protein-coupled receptor kinase subfamily of the serine⁄threonine protein kinase family. GRK4 phosphorylates the activated forms of G protein-coupled receptors.Specifications
NP_892027 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
94kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
GRK4, GPRK4 | |
GRK4 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β−glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 10μM ATP, 100 μg/mL GRKtide peptide substrate (RRREEEEESAAA) and trace [32P]- γ-ATP for 10 minutes at 30°C | |
GRK4 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.41mg/mL | |
2868, 4775 | |
GRK4 Recombinant Human Protein | |
–80°C | |
1 Each | |
GRK4 Recombinant Human Protein | |
GSK kinase family | |
12.5nmoles of phosphate transferred to GRKtide peptide substrate (RRREEEEESAAA) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2.08μg/mL | |
GST Tag | |
Kinases and inhibitors | |
90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.