missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Griseofulvin, 97%, Thermo Scientific Chemicals
14.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 217.0°C to 224.0°C |
Color | White to White-Yellow |
Loss on Drying | 1% max. (1 g, 105°C) |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Quantity | 5 g |
Merck Index | 15, 4584 |
Show More |
Safety and Handling
Product Identifier
- (+)-Griseofulvin
- Danger
- Carcinogenicity Category 2
- Reproductive toxicity Category 1A
- Respiratory sensitiser Category 1
- H360FD-May damage fertility. May damage the unborn child.
- H351-Suspected of causing cancer.
- H317-May cause an allergic skin reaction.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'